Search
Search

Piezo2| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039485.4 | NP_001034574.4 | 8 - 9 | 1994 | 62 | |
| GenBank mRNA | AK018503.1 | - | 4 - 5 | 757 | 62 | |
| AK050232.1 | - | 2 - 3 | 384 | 62 | ||
| AK053702.1 | - | 8 - 9 | 1994 | 62 | ||
| AK087529.1 | - | 8 - 9 | 1269 | 62 | ||
| HQ215521.1 | - | 8 - 9 | 1080 | 62 | ||
Set Membership: |
|