Search
Search
| Gene Symbol: Cog5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163126.1 | NP_001156598.1 | 5 - 6 | 440 | 64 | |
| GenBank mRNA | AK030656.1 | - | 5 - 6 | 441 | 64 | |
| CA316530.1 | - | 2 - 3 | 95 | 64 | ||
Set Membership: |
|