Search
Search

Klhl21| Gene Symbol: Klhl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033352.3 | NP_001028524.1 | 1 - 2 | 1073 | 72 | |
| GenBank mRNA | AK052203.1 | - | 1 - 2 | 1048 | 72 | |
| AK154493.1 | - | 1 - 2 | 1074 | 72 | ||
| AK158141.1 | - | 1 - 2 | 1049 | 72 | ||
| BC060201.1 | - | 1 - 2 | 1062 | 72 | ||
| BC141158.1 | - | 1 - 2 | 1047 | 9055770 | 72 | |
| BC141159.1 | - | 1 - 2 | 1047 | 9055771 | 72 | |
Set Membership: |
|