Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01266961_s1
          See other MGAT2 GE Assays ›
          Gene Symbol:
          Mgat2
          Gene Name:
          mannoside acetylglucosaminyltransferase 2
          Gene Aliases:
          AA407964, CDGS2, GLCNACTII, GNT-II, GNT2
          Chromosome Location:
          Chr.12: 69184158 - 69186773 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01266961_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mgat2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146035.2 NP_666147.1 1 1887 82
          GenBank mRNA AK083370.1 - 1 1844 82
          AK151964.1 - 1 1835 82
          AK152925.1 - 1 1856 82
          AK153382.1 - 1 1835 82
          AK159185.1 - 2 1761 82
          AK166037.1 - 1 1852 82
          AK171000.1 - 1 1891 82
          BC010583.1 - 1 1832 3500709 82
          BC027169.1 - 1 1832 4954047 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mgat2

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein N-linked glycosylation
          oligosaccharide biosynthetic process
          protein N-linked glycosylation via asparagine
          alpha-1,6-mannosylglycoprotein 2-beta-N-acetylglucosaminyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513230 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513224 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 BC010583.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 BC027169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 AK171000.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 AK153382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 AK159185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 BC010583.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 BC027169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 AK171000.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 AK153382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150521 AK159185.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 171569 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127527 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 BC010583.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 BC027169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 AK171000.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 AK153382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 AK159185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 BC010583.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 BC027169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 AK171000.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 AK153382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150521 AK159185.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 BC010583.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 BC027169.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 AK171000.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 AK153382.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 AK159185.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 BC010583.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 BC027169.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 AK171000.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 AK153382.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150521 AK159185.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28513253 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127527 - -
          Whole Mouse Genome G4122A A_51_P150521 BC010583.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150521 BC027169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150521 AK171000.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150521 AK153382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150521 AK159185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150521 BC010583.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150521 BC027169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150521 AK171000.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150521 AK153382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150521 AK159185.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125160 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127527 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10418423 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10418402 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513230 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28513227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28513297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10218597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10418422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28513233 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10396075 - -
          Affymetrix HT MG-430 PM Array Plate 1452037_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1426350_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10396074 - -
          Mouse Genome 430 2.0 Array 1452037_PM_at - -
          Mouse Genome 430 2.0 Array 1426350_at - -
          Mouse Genome 430 2.0 Array 1426350_PM_at - -
          Mouse Genome 430 2.0 Array 1452037_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217664_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217664_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200003749.mm.1 NM_146035.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003749.mm.1 NM_146035.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003749.mm.1 TC1200000563.mm.1 MTA TC
          Mouse Expression Set 430 1426350_PM_at - -
          Mouse Expression Set 430 1452037_at - -
          Mouse Expression Set 430 1452037_PM_at - -
          Mouse Expression Set 430 1426350_at - -
          Murine Genome U74v2 Set 95417_at - -
          Murine Genome U74v2 Set 95417_PM_at - -
          Mouse Genome 430A 2.0 Array 1452037_PM_at - -
          Mouse Genome 430A 2.0 Array 1426350_PM_at - -
          Mouse Genome 430A 2.0 Array 1452037_at - -
          Mouse Genome 430A 2.0 Array 1426350_at - -
          Affymetrix HT Mouse Genome 430A Array 1426350_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452037_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452037_at - -
          Affymetrix HT Mouse Genome 430A Array 1426350_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4760546 - -
          Illumina mouse-6 v1.0 expression beadchip 4760546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004760546 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2708279 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2708279 - -
          mouse-6 v1.1 (Illumina) scl43159.2_197-S - -
          Illumina Sentrix mouse-6.v1 bead array 4760546 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43159.2_197-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4760546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2708279 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2708279 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline