Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01267293_m1
          See other GM12789 GE Assays ›
          Gene Symbol:
          Gm12789
          Gene Name:
          predicted gene 12789
          Gene Aliases:
          OTTMUSG00000008445
          Chromosome Location:
          Chr.4: 101986840 - 101990231 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01267293_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm12789
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085520.2 NP_001078989.1 3 - 4 571 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001045.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029808 NM_001085520.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029809 NM_001085520.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34879 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029808 NM_001085520.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029809 NM_001085520.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23157773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03991454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03991458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23157733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03880112 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1457720_PM_at - -
          Mouse Genome 430 2.0 Array 1457720_at - -
          Mouse Genome 430 2.0 Array 1457720_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400007105.mm.1 NM_001085520.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007105.mm.1 NM_001085520.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007105.mm.1 TC0400001045.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400007110.mm.1 NM_001085520.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007110.mm.1 NM_001085520.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007110.mm.1 TC0400001045.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400007109.mm.1 NM_001085520.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007109.mm.1 NM_001085520.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007109.mm.1 TC0400001045.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400007106.mm.1 NM_001085520.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007106.mm.1 NM_001085520.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007106.mm.1 TC0400001045.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400007104.mm.1 NM_001085520.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007104.mm.1 NM_001085520.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007104.mm.1 TC0400001045.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400007103.mm.1 NM_001085520.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007103.mm.1 NM_001085520.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007103.mm.1 TC0400001045.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400007108.mm.1 NM_001085520.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007108.mm.1 NM_001085520.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007108.mm.1 TC0400001045.mm.1 MTA TC
          Mouse Expression Set 430 1457720_at - -
          Mouse Expression Set 430 1457720_PM_at - -
          Murine Genome U74v2 Set 135156_PM_at - -
          Murine Genome U74v2 Set 135156_at - -
          Affymetrix HT Mouse Genome 430B Array 1457720_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457720_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 703455 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline