Search
Search
| Gene Symbol: Whrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008791.2 | NP_001008791.1 | 2 - 3 | 1602 | 71 | |
| NM_001008792.2 | NP_001008792.1 | 2 - 3 | 1602 | 71 | ||
| NM_001008793.2 | NP_001008793.1 | 2 - 3 | 1602 | 71 | ||
| NM_001276371.1 | NP_001263300.1 | 2 - 3 | 1602 | 71 | ||
| NM_028640.2 | NP_082916.1 | 2 - 3 | 1602 | 71 | ||
| GenBank mRNA | AK122523.1 | - | 2 - 3 | 733 | 71 | |
| AK157955.1 | - | 2 - 3 | 1603 | 71 | ||
| AY739114.1 | - | 2 - 3 | 1562 | 71 | ||
| AY739115.1 | - | 2 - 3 | 1562 | 71 | ||
| AY739116.1 | - | 2 - 3 | 1562 | 71 | ||
| AY739117.1 | - | 2 - 3 | 1562 | 71 | ||
| BC150757.1 | - | 2 - 3 | 959 | 9087668 | 71 | |
| HQ148552.1 | - | 2 - 3 | 844 | 71 | ||
| HQ148553.1 | - | 2 - 3 | 844 | 71 | ||
Set Membership: |
|