Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01267666_sH
          See other IFNA15 GE Assays ›
          Gene Symbol:
          Ifna15
          Gene Name:
          interferon alpha 15
          Gene Aliases:
          Gm12597, Ifnaa, MuIFN-alpha-A, OTTMUSG00000007655
          Chromosome Location:
          Chr.4: 88557673 - 88558245 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01267666_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifna15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206870.1 NP_996753.1 1 237 93
          GenBank mRNA BC116216.2 - 1 237 40091189 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003002.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifna15

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          cytokine activity
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04019162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03784811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03569351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03710844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04045748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03478864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03310331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03491438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514290 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505879 - -
          Affymetrix Mouse Gene 1.1 ST Array 10514289 - -
          Affymetrix Mouse Clariom S Array PSR0400024294.mm.1 TC0400003002.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024294.mm.1 NM_206870.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024294.mm.1 NM_206870.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024295.mm.1 TC0400003002.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024295.mm.1 NM_206870.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024295.mm.1 NM_206870.1 Refseq
          Mouse Expression Set 430 1422404_PM_x_at - -
          Mouse Expression Set 430 1422403_PM_at - -
          Mouse Expression Set 430 1422403_at - -
          Mouse Expression Set 430 1450593_PM_at - -
          Mouse Expression Set 430 1450593_at - -
          Mouse Expression Set 430 1422404_x_at - -
          Murine Genome U74v2 Set 102149_PM_f_at - -
          Murine Genome U74v2 Set 102150_f_at - -
          Murine Genome U74v2 Set 102149_f_at - -
          Murine Genome U74v2 Set 102150_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1450593_at - -
          Mouse Genome 430A 2.0 Array 1422403_at - -
          Mouse Genome 430A 2.0 Array 1422404_x_at - -
          Mouse Genome 430A 2.0 Array 1450593_PM_at - -
          Mouse Genome 430A 2.0 Array 1422404_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1422403_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 416068 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104540735 - -
          Illumina mouse-6 v1.0 expression beadchip 104540735 - -
          Illumina mouse-6 v1.0 expression beadchip 610026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2655985 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2655985 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215054 - -
          mouse-6 v1.1 (Illumina) scl0242517.1_41-S - -
          mouse-6 v1.1 (Illumina) GI_46047407-S - -
          Illumina Sentrix mouse-6.v1 bead array 610026 - -
          Illumina Sentrix mouse-6.v1 bead array 104540735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0242517.1_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104540735 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2655985 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2655985 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215054 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline