Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01267981_s1
          See other C5AR2 GE Assays ›
          Gene Symbol:
          C5ar2
          Gene Name:
          complement component 5a receptor 2
          Gene Aliases:
          C5L2, E030029A11Rik, Gpr77
          Chromosome Location:
          Chr.7: 16234585 - 16244154 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01267981_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C5ar2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146005.1 NP_001139477.1 2 995 80
          NM_176912.4 NP_795886.2 2 1088 80
          GenBank mRNA AK053187.1 - 2 1089 80
          AK160050.1 - 2 996 80
          BC137941.1 - 2 977 8860960 80
          BC145834.1 - 2 977 40131137 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C5ar2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          inflammatory response
          signal transduction
          G-protein coupled receptor signaling pathway
          negative regulation of tumor necrosis factor production
          positive regulation of epithelial cell proliferation
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of neutrophil chemotaxis
          negative regulation of interleukin-6 secretion
          regulation of interleukin-8 secretion
          signal transducer activity
          G-protein coupled receptor activity
          C5a anaphylatoxin receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106106 BY540170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106106 AK053187.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106106 AK160050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106106 NM_001146005.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106106 NM_176912.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970788 BY540170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970788 AK053187.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970788 AK160050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970788 NM_001146005.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970788 NM_176912.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16376 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147218 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970788 BY540170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970788 AK053187.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970788 AK160050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970788 NM_001146005.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970788 NM_176912.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106106 BY540170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106106 AK053187.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106106 AK160050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106106 NM_001146005.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106106 NM_176912.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169696 BY540170.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169696 AK053187.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169696 AK160050.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169696 NM_001146005.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169696 NM_176912.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24980470 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06129035 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147218 - -
          Whole Mouse Genome G4122A A_51_P169696 BY540170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169696 AK053187.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169696 AK160050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169696 NM_001146005.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169696 NM_176912.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131313 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24980423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24980459 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560241 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560240 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560237 - -
          Affymetrix Mouse Clariom S Array PSR0700021307.mm.1 NM_001146005.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021307.mm.1 NM_001146005.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021307.mm.1 NM_176912.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021307.mm.1 NM_176912.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021307.mm.1 TC0700002421.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021310.mm.1 NM_001146005.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021310.mm.1 NM_001146005.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021310.mm.1 NM_176912.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021310.mm.1 NM_176912.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021310.mm.1 TC0700002421.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021311.mm.1 NM_001146005.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021311.mm.1 NM_001146005.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021311.mm.1 TC0700002421.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021313.mm.1 NM_176912.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021313.mm.1 NM_176912.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021313.mm.1 TC0700002421.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021308.mm.1 NM_001146005.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021308.mm.1 NM_001146005.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021308.mm.1 NM_176912.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021308.mm.1 NM_176912.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021308.mm.1 TC0700002421.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021309.mm.1 NM_001146005.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021309.mm.1 NM_001146005.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021309.mm.1 NM_176912.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021309.mm.1 NM_176912.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021309.mm.1 TC0700002421.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 424095 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4200022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200022 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2671707 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2671707 - -
          mouse-6 v1.1 (Illumina) scl31726.4_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200022 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31726.4_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2671707 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2671707 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline