Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268112_m1
          See other GM10451 GE Assays ›
          Gene Symbol:
          Gm10451
          Gene Name:
          predicted gene 10451
          Gene Aliases:
          ENSMUSG00000073000
          Chromosome Location:
          Chr.12: 76444497 - 76448698 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268112_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10451
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028520.1 1 - 2 169 102
          GenBank mRNA AK136613.1 - 108 102

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK136613.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2167728 NR_028520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P11216 AK136613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2167728 NR_028520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P11216 AK136613.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07671423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10123921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09801412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10323279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09766745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09766748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09966271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09860097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10314367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10151704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10109062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10123925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09866513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28554001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1445468_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457869_PM_at - -
          Mouse Genome 430 2.0 Array 1457869_at - -
          Mouse Genome 430 2.0 Array 1457869_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200004842.mm.1 NR_028520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004842.mm.1 NR_028520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004842.mm.1 TC1200000681.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200004841.mm.1 NR_028520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004841.mm.1 NR_028520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004841.mm.1 TC1200000681.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200004839.mm.1 NR_028520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004839.mm.1 NR_028520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004839.mm.1 TC1200000681.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200004840.mm.1 NR_028520.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004840.mm.1 NR_028520.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004840.mm.1 TC1200000681.mm.1 MTA TC
          Mouse Expression Set 430 1457869_at - -
          Mouse Expression Set 430 1457869_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457869_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457869_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 658411 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline