Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268529_m1
          See other DYNAP GE Assays ›
          Gene Symbol:
          Dynap
          Gene Name:
          dynactin associated protein
          Gene Aliases:
          2310002L13Rik
          Chromosome Location:
          Chr.18: 70240429 - 70244584 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268529_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dynap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029346.1 NP_083622.1 2 - 3 347 100
          GenBank mRNA AK009098.1 - 2 - 3 162 100
          HQ404309.1 - 2 - 3 260 100
          HQ404310.1 - 2 - 3 260 100
          HQ404311.1 - 2 - 3 260 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Process(es)

          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114651 NM_029346.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114651 AK009098.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123231 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114651 NM_029346.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114651 AK009098.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123231 - -
          Whole Mouse Genome G4122A A_51_P179293 NM_029346.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P179293 AK009098.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117572 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121233 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14071790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14061328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14061351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31808636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13921461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453275_PM_at - -
          Mouse Genome 430 2.0 Array 1453275_PM_at - -
          Mouse Genome 430 2.0 Array 1453275_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75577_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75577_at - -
          Affymetrix Mouse Clariom S Array PSR1800011250.mm.1 NM_029346.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011250.mm.1 NM_029346.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011250.mm.1 TC1800001537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011251.mm.1 NM_029346.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011251.mm.1 NM_029346.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011251.mm.1 TC1800001537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011252.mm.1 NM_029346.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011252.mm.1 NM_029346.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011252.mm.1 TC1800001537.mm.1 MTA TC
          Mouse Expression Set 430 1453275_PM_at - -
          Mouse Expression Set 430 1453275_at - -
          Murine Genome U74v2 Set 170916_i_at - -
          Murine Genome U74v2 Set 167419_PM_f_at - -
          Murine Genome U74v2 Set 170916_PM_i_at - -
          Murine Genome U74v2 Set 167419_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1453275_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453275_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 504324 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5860092 - -
          Illumina mouse-6 v1.0 expression beadchip 5860092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2742813 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2742813 - -
          mouse-6 v1.1 (Illumina) scl51301.3.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 5860092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51301.3.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860092 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2742813 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0