Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268569_m1
          See other KCNMA1 GE Assays ›
          Gene Symbol:
          Kcnma1
          Gene Name:
          potassium large conductance calcium-activated channel, subfamily M, alpha member 1
          Gene Aliases:
          5730414M22Rik, BKCa, MaxiK, Slo, Slo1, mSlo, mSlo1
          Chromosome Location:
          Chr.14: 23292291 - 24004951 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268569_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnma1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253358.1 NP_001240287.1 10 - 11 1410 86
          NM_001253359.1 NP_001240288.1 10 - 11 1410 86
          NM_001253360.1 NP_001240289.1 10 - 11 1410 86
          NM_001253361.1 NP_001240290.1 10 - 11 1410 86
          NM_001253362.1 NP_001240291.1 10 - 11 1410 86
          NM_001253363.1 NP_001240292.1 10 - 11 1410 86
          NM_001253364.1 NP_001240293.1 10 - 11 1410 86
          NM_001253365.1 NP_001240294.1 10 - 11 1410 86
          NM_001253366.1 NP_001240295.1 10 - 11 1410 86
          NM_001253367.1 NP_001240296.1 10 - 11 1410 86
          NM_001253368.1 NP_001240297.1 10 - 11 1410 86
          NM_001253369.1 NP_001240298.1 10 - 11 1410 86
          NM_001253370.1 NP_001240299.1 10 - 11 1410 86
          NM_001253371.1 NP_001240300.1 10 - 11 1410 86
          NM_001253372.1 NP_001240301.1 10 - 11 1410 86
          NM_001253373.1 NP_001240302.1 10 - 11 1410 86
          NM_001253374.1 NP_001240303.1 9 - 10 1248 86
          NM_001253375.1 NP_001240304.1 10 - 11 1410 86
          NM_001253376.1 NP_001240305.1 10 - 11 1410 86
          NM_001253377.1 NP_001240306.1 10 - 11 1410 86
          NM_001253378.1 NP_001240307.1 10 - 11 1410 86
          NM_010610.3 NP_034740.2 10 - 11 1410 86
          GenBank mRNA AF156674.1 - 10 - 11 1135 86
          AF465244.1 - 10 - 11 1135 86
          AK142894.1 - 10 - 11 1354 86
          AK149333.1 - 6 - 7 955 86
          AK166436.1 - 1067 86
          AK166536.1 - 9 - 10 1023 86
          BC065068.1 - 10 - 11 1410 86
          BC128331.1 - 10 - 11 1132 86
          FJ872117.1 - 10 - 11 1135 86
          HQ221743.1 - 10 - 11 1135 86
          HQ221744.1 - 9 - 10 973 86
          HQ221745.1 - 10 - 11 1135 86
          HQ221746.1 - 10 - 11 1135 86
          HQ221747.1 - 10 - 11 1135 86
          HQ221748.1 - 10 - 11 1135 86
          HQ221749.1 - 10 - 11 1135 86
          HQ221750.1 - 10 - 11 1135 86
          HQ221751.1 - 10 - 11 1135 86
          HQ221752.1 - 10 - 11 1135 86
          HQ221753.1 - 10 - 11 1135 86
          HQ221754.1 - 10 - 11 1135 86
          HQ221755.1 - 10 - 11 1135 86
          HQ221756.1 - 10 - 11 1135 86
          HQ221757.1 - 10 - 11 1135 86
          HQ221759.1 - 10 - 11 1135 86
          HQ221760.1 - 10 - 11 1135 86
          HQ221761.1 - 10 - 11 1135 86
          HQ221762.1 - 10 - 11 1135 86
          HQ221763.1 - 10 - 11 1135 86
          HQ529469.1 - 11 - 12 1227 86
          HQ529471.1 - 10 - 11 1135 86
          HQ529475.1 - 10 - 11 1135 86
          HQ529477.1 - 10 - 11 1135 86
          L16912.1 - 10 - 11 1216 86
          U09383.1 - 10 - 11 2075 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001567.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnma1

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          transport
          ion transport
          potassium ion transport
          response to osmotic stress
          chemical synaptic transmission
          sensory perception of sound
          circadian rhythm
          adult walking behavior
          response to pH
          neuronal action potential
          cellular potassium ion homeostasis
          response to corticosteroid
          regulation of aldosterone metabolic process
          response to carbon monoxide
          regulation of ion transmembrane transport
          vasodilation
          regulation of vasodilation
          regulation of membrane potential
          auditory receptor cell differentiation
          positive regulation of apoptotic process
          response to estrogen
          locomotor rhythm
          negative regulation of cell volume
          saliva secretion
          cell maturation
          neuromuscular process controlling balance
          protein homooligomerization
          protein homotetramerization
          response to calcium ion
          transmembrane transport
          micturition
          eye blink reflex
          smooth muscle contraction involved in micturition
          relaxation of vascular smooth muscle
          potassium ion transmembrane transport
          actin binding
          ion channel activity
          voltage-gated ion channel activity
          voltage-gated potassium channel activity
          potassium channel activity
          protein binding
          calcium-activated potassium channel activity
          protein complex binding
          protein homodimerization activity
          metal ion binding
          large conductance calcium-activated potassium channel activity

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 924022 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0