Search
Search

Tmem87a| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110496.1 | NP_001103966.1 | 15 - 16 | 1476 | 64 | |
| NM_001110497.1 | NP_001103967.1 | 15 - 16 | 1476 | 64 | ||
| NM_173734.3 | NP_776095.2 | 15 - 16 | 1479 | 64 | ||
| GenBank mRNA | AK044591.1 | - | 15 - 16 | 1430 | 64 | |
| AK086738.1 | - | 15 - 16 | 1426 | 64 | ||
| AK134268.1 | - | 15 - 16 | 1442 | 64 | ||
| BC004047.1 | - | 15 - 16 | 1425 | 64 | ||
| BC024676.1 | - | 5 - 6 | 427 | 64 | ||
| BC027354.1 | - | 15 - 16 | 1452 | 4010477 | 64 | |
Set Membership: |
|