| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10476131 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10476132 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1430422_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10476130 |
- |
- |
| Mouse Genome 430 2.0 Array |
1430422_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1430422_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
75656_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
75656_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0200015698.mm.1 |
NM_029375.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015698.mm.1 |
NM_029375.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015698.mm.1 |
TC0200001980.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015699.mm.1 |
NM_001163483.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015699.mm.1 |
NM_001163483.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015699.mm.1 |
TC0200001980.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015701.mm.1 |
NM_001163483.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015701.mm.1 |
NM_001163483.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015701.mm.1 |
NM_029375.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015701.mm.1 |
NM_029375.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015701.mm.1 |
TC0200001980.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015700.mm.1 |
NM_001163483.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015700.mm.1 |
NM_001163483.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015700.mm.1 |
NM_029375.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015700.mm.1 |
NM_029375.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015700.mm.1 |
TC0200001980.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1430422_PM_at |
- |
- |
| Mouse Expression Set 430 |
1430422_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1430422_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1430422_at |
- |
- |