Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268692_m1
          See other AA467197 GE Assays ›
          Gene Symbol:
          AA467197
          Gene Name:
          expressed sequence AA467197
          Gene Aliases:
          Nmes1, mir-147
          Chromosome Location:
          Chr.2: 122636971 - 122641087 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268692_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AA467197
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004174.1 NP_001004174.1 4 - 5 258 65
          GenBank mRNA AK141297.1 - 3 - 4 361 65
          AK150561.1 - 4 - 5 248 65
          BC049633.1 - 4 - 5 258 6774154 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001854.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AA467197

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587204 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587209 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587208 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587210 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587205 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587203 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587202 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587211 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137049 AK141297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137049 AK150561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137049 BC049633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137049 NM_001004174.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143442 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163751 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137049 AK141297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137049 AK150561.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137049 BC049633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137049 NM_001004174.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21587244 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163751 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163751 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01943831 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02140053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01943839 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01943849 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01929963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587210 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587203 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587204 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587205 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587209 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587211 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587202 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587208 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21587212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21587223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01155443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01929962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01241815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01155447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01900164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02184191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01300242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01298329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01943844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02140055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01300235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01155455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01185333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01300229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01298328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01298330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01241817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01900162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01249491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01298332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02048782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01300241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21587264 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10475519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475520 - -
          Affymetrix HT MG-430 PM Array Plate 1434046_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10475517 - -
          Mouse Genome 430 2.0 Array 1434046_PM_at - -
          Mouse Genome 430 2.0 Array 1434046_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433470_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433470_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200014441.mm.1 NM_001004174.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014441.mm.1 NM_001004174.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014441.mm.1 TC0200001854.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014438.mm.1 NM_001004174.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014438.mm.1 NM_001004174.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014438.mm.1 TC0200001854.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014439.mm.1 NM_001004174.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014439.mm.1 NM_001004174.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014439.mm.1 NR_030547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014439.mm.1 NR_030547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014439.mm.1 TC0200001854.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014440.mm.1 NM_001004174.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014440.mm.1 NM_001004174.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014440.mm.1 TC0200001854.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014437.mm.1 NM_001004174.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014437.mm.1 NM_001004174.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014437.mm.1 TC0200001854.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014436.mm.1 NM_001004174.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014436.mm.1 NM_001004174.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014436.mm.1 TC0200001854.mm.1 MTA TC
          Mouse Expression Set 430 1434046_at - -
          Mouse Expression Set 430 1434046_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434046_at - -
          Affymetrix HT Mouse Genome 430B Array 1434046_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 576673 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160750 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160750 - -
          mouse-6 v1.1 (Illumina) GI_51921340-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160750 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160750 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_51921340-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline