Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268754_m1
          See other CCL20 GE Assays ›
          Gene Symbol:
          Ccl20
          Gene Name:
          chemokine (C-C motif) ligand 20
          Gene Aliases:
          CKb4, LARC, MIP-3A, MIP-3[a], MIP3A, ST38, Scya20, exodus-1
          Chromosome Location:
          Chr.1: 83116744 - 83119167 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268754_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccl20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159738.1 NP_001153210.1 3 - 4 330 102
          NM_016960.2 NP_058656.1 3 - 4 333 102
          GenBank mRNA AB015136.1 - 3 - 4 317 102
          AF053313.1 - 3 - 4 309 102
          BC028504.1 - 3 - 4 303 1380543 102
          BX525886.1 - 3 - 4 311 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000718.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccl20

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          inflammatory response
          immune response
          positive regulation of interleukin-1 alpha biosynthetic process
          positive regulation of nitric-oxide synthase biosynthetic process
          cell chemotaxis
          positive regulation of T cell migration
          cytokine activity
          protein binding
          chemokine activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175885 AK079862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175885 BC028504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175885 NM_001159738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175885 NM_016960.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175885 AB015136.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841707 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100947 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2175885 AK079862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2175885 BC028504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2175885 NM_001159738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2175885 NM_016960.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2175885 AB015136.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408595 AK079862.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408595 BC028504.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408595 NM_001159738.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408595 NM_016960.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408595 AB015136.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20396693 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100947 - -
          Whole Mouse Genome G4122A A_51_P408595 AK079862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408595 BC028504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408595 NM_001159738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408595 NM_016960.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408595 AB015136.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108309 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100947 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00131528 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00491075 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00491094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00491086 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00131527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20396668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00491086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00504997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00505011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00131528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00504998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00885219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00927025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396683 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10347890 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347893 - -
          Affymetrix HT MG-430 PM Array Plate 1422029_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10347888 - -
          Mouse Genome 430 2.0 Array 1422029_PM_at - -
          Mouse Genome 430 2.0 Array 1422029_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20297_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20297_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100006103.mm.1 NM_001159738.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100006103.mm.1 NM_001159738.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100006103.mm.1 NM_016960.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100006103.mm.1 NM_016960.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100006103.mm.1 TC0100000718.mm.1 MTA TC
          Mouse Expression Set 430 1422029_at - -
          Mouse Expression Set 430 1422029_PM_at - -
          Mouse Genome 430A 2.0 Array 1422029_PM_at - -
          Mouse Genome 430A 2.0 Array 1422029_at - -
          Affymetrix HT Mouse Genome 430A Array 1422029_at - -
          Affymetrix HT Mouse Genome 430A Array 1422029_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105130300 - -
          Illumina mouse-6 v1.0 expression beadchip 105130300 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2639936 - -
          mouse-6 v1.1 (Illumina) scl17735.3_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 105130300 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105130300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2781178 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218692 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2781178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0