| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200012616.mm.1 |
NM_001145898.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012616.mm.1 |
NM_001145898.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012616.mm.1 |
NM_207264.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012616.mm.1 |
NM_207264.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012616.mm.1 |
TC0200001700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012617.mm.1 |
NM_001145898.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012617.mm.1 |
NM_001145898.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012617.mm.1 |
NM_207264.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012617.mm.1 |
NM_207264.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012617.mm.1 |
TC0200001700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012615.mm.1 |
NM_001145898.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012615.mm.1 |
NM_001145898.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012615.mm.1 |
NM_207264.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012615.mm.1 |
NM_207264.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012615.mm.1 |
TC0200001700.mm.1 |
MTA TC |