Search
Search

Tex36| Gene Symbol: Tex36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028654.1 | NP_082930.1 | 1 - 2 | 153 | 83 | |
| GenBank mRNA | AK015080.1 | - | 1 - 2 | 198 | 83 | |
| BC100509.1 | - | 1 - 2 | 153 | 30918228 | 83 | |
Set Membership: |
|