Search
Search
| Gene Symbol: Uros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA272198.1 | - | 4 - 5 | 423 | 70 | |
| AK135314.1 | - | 5 - 6 | 479 | 70 | ||
| AK145902.1 | - | 5 - 6 | 529 | 70 | ||
| AK148116.1 | - | 5 - 6 | 666 | 70 | ||
| AK164297.1 | - | 5 - 6 | 444 | 70 | ||
| BC068161.1 | - | 5 - 6 | 472 | 30358504 | 70 | |
| CO044764.1 | - | 2 - 3 | 70 | |||
| U04439.1 | - | 4 - 5 | 393 | 70 | ||
| U16216.1 | - | 5 - 6 | 534 | 70 | ||
| U18867.2 | - | 5 - 6 | 538 | 70 | ||
Set Membership: |
|