Search
Search

St6gal2| Gene Symbol: St6gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172829.2 | NP_766417.1 | 3 - 4 | 1184 | 105 | |
| GenBank mRNA | AK082566.1 | - | 3 - 4 | 1185 | 105 | |
| AK129462.1 | - | 1 - 2 | 116 | 105 | ||
| BC125586.1 | - | 2 - 3 | 1054 | 40130101 | 105 | |
| BC138142.1 | - | 2 - 3 | 1054 | 8861161 | 105 | |
Set Membership: |
|