Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01269572_s1
          See other OLFR645 GE Assays ›
          Gene Symbol:
          Olfr645
          Gene Name:
          olfactory receptor 645
          Gene Aliases:
          MOR13-5
          Chromosome Location:
          Chr.7: 104084131 - 104085078 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01269572_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr645
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207144.1 NP_997027.1 1 867 97
          GenBank mRNA BC132009.1 - 1 965 40130286 97
          BC132011.1 - 1 966 40130288 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr645

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976554 BC132009.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976554 BC132011.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976554 NM_207144.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115170 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976554 BC132009.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976554 BC132011.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976554 NM_207144.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483103 BC132009.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483103 BC132011.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483103 NM_207144.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386596 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115170 - -
          Whole Mouse Genome G4122A A_51_P483103 BC132009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483103 BC132011.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483103 NM_207144.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115170 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06407669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06407694 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06407674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06407708 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06407693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06407703 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06407687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06407696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06407691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06014211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06227018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06407668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06407709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06175980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06014205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06407707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06507092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386583 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566303 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566302 - -
          Affymetrix Mouse Clariom S Array PSR0700032838.mm.1 TC0700003925.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032837.mm.1 NM_207144.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032837.mm.1 NM_207144.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032837.mm.1 TC0700003925.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032836.mm.1 TC0700003925.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3710692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2738793 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2738793 - -
          mouse-6 v1.1 (Illumina) scl0258247.1_100-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258247.1_100-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238709 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238709 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline