Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01269910_m1
          See other SLC35B2 GE Assays ›
          Gene Symbol:
          Slc35b2
          Gene Name:
          solute carrier family 35, member B2
          Gene Aliases:
          1110003M08Rik, AA407792, AI842275, PAPST1, Slc35b1
          Chromosome Location:
          Chr.17: 45563879 - 45567671 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01269910_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc35b2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028662.2 NP_082938.1 2 - 3 777 67
          GenBank mRNA AF414190.1 - 3 - 4 450 67
          AK090329.1 - 2 - 3 966 67
          AK144403.1 - 3 - 4 473 67
          AK152362.1 - 3 - 4 451 67
          AK154868.1 - 3 - 4 436 67
          AK155551.1 - 3 - 4 483 67
          AK167343.1 - 2 - 3 778 67
          BC025875.1 - 3 - 4 449 5137689 67
          BC036992.1 - 3 - 4 457 5003855 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000865.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          3'-phosphoadenosine 5'-phosphosulfate transport
          transmembrane transport
          3'-phospho-5'-adenylyl sulfate transmembrane transport
          signal transducer activity
          3'-phosphoadenosine 5'-phosphosulfate transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31201836 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31201839 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31201835 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31201834 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086094 AK212091.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086094 AK214463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086094 BC025875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086094 BC036992.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086094 AK154868.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776829 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155816 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105348 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086094 AK212091.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086094 AK214463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086094 BC025875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086094 BC036992.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086094 AK154868.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105348 - -
          Whole Mouse Genome G4122A A_52_P427564 AK212091.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P427564 AK214463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P427564 BC025875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P427564 BC036992.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P427564 AK154868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174384 AK212100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174384 AK214463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174384 BC025875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174384 BC036992.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174384 AK154868.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105348 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13156855 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13359840 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13193658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31201834 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31201839 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31201835 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31201836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31201795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13485988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13485996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13115217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13359840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13115234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13485984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31201854 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445427 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445425 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445426 - -
          Affymetrix HT MG-430 PM Array Plate 1429007_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423927_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445422 - -
          Mouse Genome 430 2.0 Array 1423927_at - -
          Mouse Genome 430 2.0 Array 1429007_at - -
          Mouse Genome 430 2.0 Array 1429007_PM_at - -
          Mouse Genome 430 2.0 Array 1423927_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73836_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73836_at - -
          Affymetrix Mouse Clariom S Array PSR1700008243.mm.1 NM_028662.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008243.mm.1 NM_028662.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008243.mm.1 TC1700000865.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008244.mm.1 NM_028662.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008244.mm.1 NM_028662.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008244.mm.1 TC1700000865.mm.1 MTA TC
          Mouse Expression Set 430 1423927_at - -
          Mouse Expression Set 430 1423927_PM_at - -
          Mouse Expression Set 430 1429007_at - -
          Mouse Expression Set 430 1429007_PM_at - -
          Murine Genome U74v2 Set 130444_at - -
          Murine Genome U74v2 Set 130444_PM_at - -
          Mouse Genome 430A 2.0 Array 1423927_at - -
          Mouse Genome 430A 2.0 Array 1423927_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423927_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423927_at - -
          Affymetrix HT Mouse Genome 430B Array 1429007_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429007_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 445066 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246740 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2630366 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246740 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2630366 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline