Search
Search

Fgfr2| Gene Symbol: Fgfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010207.2 | NP_034337.2 | 14 - 15 | 3209 | 76 | |
| NM_201601.2 | NP_963895.2 | 12 - 13 | 2867 | 76 | ||
| GenBank mRNA | BC065069.1 | - | 12 - 13 | 2289 | 76 | |
| BC072640.1 | - | 13 - 14 | 2501 | 76 | ||
| BC151201.1 | - | 12 - 13 | 2069 | 9088089 | 76 | |
| BC172174.1 | - | 13 - 14 | 2150 | 9054844 | 76 | |
| EF143322.1 | - | 12 - 13 | 1721 | 76 | ||
| EF143323.1 | - | 13 - 14 | 1988 | 76 | ||
| EF143324.1 | - | 11 - 12 | 1643 | 76 | ||
| EF143325.1 | - | 12 - 13 | 1715 | 76 | ||
| EF143326.1 | - | 12 - 13 | 1721 | 76 | ||
| EF143327.1 | - | 11 - 12 | 1637 | 76 | ||
| EF143328.1 | - | 13 - 14 | 1982 | 76 | ||
| EF143329.1 | - | 13 - 14 | 1988 | 76 | ||
| EF143330.1 | - | 11 - 12 | 1532 | 76 | ||
| EF143331.1 | - | 11 - 12 | 1637 | 76 | ||
| EF143332.1 | - | 13 - 14 | 1866 | 76 | ||
| EF143333.1 | - | 9 - 10 | 1346 | 76 | ||
| EF143334.1 | - | 11 - 12 | 1634 | 76 | ||
| EF143335.1 | - | 13 - 14 | 1979 | 76 | ||
| EF143336.1 | - | 11 - 12 | 1643 | 76 | ||
| EF143337.1 | - | 9 - 10 | 1349 | 76 | ||
| EF143338.1 | - | 13 - 14 | 1985 | 76 | ||
| EF143339.1 | - | 11 - 12 | 1634 | 76 | ||
| EF143340.1 | - | 11 - 12 | 1640 | 76 | ||
| M23362.1 | - | 4 - 5 | 557 | 76 | ||
| M63503.1 | - | 12 - 13 | 2318 | 76 | ||
| M86441.1 | - | 14 - 15 | 2591 | 76 | ||
| X55441.1 | - | 14 - 15 | 2639 | 76 | ||
Set Membership: |
|