Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01271171_g1
          See other BRD2 GE Assays ›
          Gene Symbol:
          Brd2
          Gene Name:
          bromodomain containing 2
          Gene Aliases:
          AW228947, D17H6S113E, Frg-1, Fsrg-1, Fsrg1, Nat, Ring3, Rnf3, mKIAA4005
          Chromosome Location:
          Chr.17: 34112019 - 34122607 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01271171_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Brd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204973.1 NP_001191902.1 9 - 10 3283 63
          NM_010238.3 NP_034368.2 9 - 10 3102 63
          NR_037970.1 9 - 10 2929 63
          GenBank mRNA AB010246.1 - 8 - 9 1607 63
          AB010247.1 - 8 - 9 1576 63
          AB010248.1 - 7 - 8 1473 63
          AB212273.1 - 7 - 8 1437 63
          AF045462.1 - 8 - 9 2248 63
          AK158970.1 - 9 - 10 3103 63
          AK168423.1 - 9 - 10 2361 63
          AK168525.1 - 9 - 10 2974 63
          AK188169.1 - 2 - 3 369 63
          AK220444.1 - 8 - 9 2250 63
          BC046809.1 - 9 - 10 1964 63
          BC085251.1 - 10 - 11 3125 63
          BC138655.1 - 8 - 9 1876 8861675 63
          BC138656.1 - 8 - 9 1876 8861676 63
          KJ650222.1 - 8 - 9 1568 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Brd2

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          chromatin modification
          chromatin binding
          lysine-acetylated histone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700017724.mm.1 NM_001204973.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017724.mm.1 NM_001204973.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017724.mm.1 NM_010238.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017724.mm.1 NM_010238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017724.mm.1 NR_037970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017724.mm.1 NR_037970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017724.mm.1 TC1700001898.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017720.mm.1 NM_001204973.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017720.mm.1 NM_001204973.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017720.mm.1 NM_010238.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017720.mm.1 NM_010238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017720.mm.1 NR_037970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017720.mm.1 NR_037970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017720.mm.1 TC1700001898.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017723.mm.1 NM_001204973.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017723.mm.1 NM_001204973.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017723.mm.1 NM_010238.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017723.mm.1 NM_010238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017723.mm.1 NR_037970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017723.mm.1 NR_037970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017723.mm.1 TC1700001898.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline