Search
Search

Syndig1| Gene Symbol: Syndig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085521.2 | NP_001078990.1 | 2 - 3 | 695 | 62 | |
| GenBank mRNA | BC147352.1 | - | 1 - 2 | 496 | 9056389 | 62 |
| BC147353.1 | - | 1 - 2 | 496 | 9056409 | 62 | |
| BC147660.1 | - | 1 - 2 | 496 | 9087449 | 62 | |
| BC147661.1 | - | 1 - 2 | 496 | 9087450 | 62 | |
Set Membership: |
|