Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01271729_m1
          See other KBTBD4 GE Assays ›
          Gene Symbol:
          Kbtbd4
          Gene Name:
          kelch repeat and BTB (POZ) domain containing 4
          Gene Aliases:
          2510026C23Rik
          Chromosome Location:
          Chr.2: 90904724 - 90910561 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01271729_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kbtbd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025991.3 NP_080267.1 3 - 4 846 81
          GenBank mRNA AK010998.1 - 3 - 4 847 81
          AK014373.1 - 3 - 4 772 81
          BC025103.1 - 3 - 4 751 4481431 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001359.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kbtbd4

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21419567 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21419566 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21419569 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21419568 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002117 BC025103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002117 AK014373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002117 NM_025991.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002117 AK010998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002117 AK168416.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831906 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100252 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002117 BC025103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002117 AK014373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002117 NM_025991.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002117 AK010998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002117 AK168416.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21419569 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00429039 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100252 - -
          Whole Mouse Genome G4122A A_51_P195439 BC025103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195439 AK014373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195439 NM_025991.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195439 AK010998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195439 AK168416.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124386 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100252 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01927643 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01927654 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01927640 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01927653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21419566 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21419569 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21419567 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21419568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21419611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01631910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01601793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01631902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01927653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21419610 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473755 - -
          Affymetrix HT MG-430 PM Array Plate 1417922_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10473754 - -
          Mouse Genome 430 2.0 Array 1417922_at - -
          Mouse Genome 430 2.0 Array 1445621_PM_at - -
          Mouse Genome 430 2.0 Array 1445621_at - -
          Mouse Genome 430 2.0 Array 1417922_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67136_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67136_at - -
          Affymetrix Mouse Clariom S Array PSR0200010401.mm.1 NM_025991.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010401.mm.1 NM_025991.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010401.mm.1 TC0200001359.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010403.mm.1 NM_025991.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010403.mm.1 NM_025991.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010403.mm.1 TC0200001359.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010400.mm.1 NM_025991.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010400.mm.1 NM_025991.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010400.mm.1 TC0200001359.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010402.mm.1 NM_025991.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010402.mm.1 NM_025991.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010402.mm.1 TC0200001359.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010404.mm.1 NM_025991.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010404.mm.1 NM_025991.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010404.mm.1 TC0200001359.mm.1 MTA TC
          Mouse Expression Set 430 1417922_at - -
          Mouse Expression Set 430 1417922_PM_at - -
          Mouse Genome 430A 2.0 Array 1417922_PM_at - -
          Mouse Genome 430A 2.0 Array 1417922_at - -
          Affymetrix HT Mouse Genome 430A Array 1417922_at - -
          Affymetrix HT Mouse Genome 430A Array 1417922_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 575685 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3840093 - -
          Illumina mouse-6 v1.0 expression beadchip 70465 - -
          Illumina mouse-6 v1.0 expression beadchip 3840093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070465 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731145 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243768 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243768 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731145 - -
          mouse-6 v1.1 (Illumina) scl0003083.1_1-S - -
          mouse-6 v1.1 (Illumina) scl20651.5_100-S - -
          Illumina Sentrix mouse-6.v1 bead array 70465 - -
          Illumina Sentrix mouse-6.v1 bead array 3840093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20651.5_100-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003083.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840093 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70465 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2806290 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2806291 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2806291 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2806290 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003083.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0