Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01271763_m1
          See other MADD GE Assays ›
          Gene Symbol:
          Madd
          Gene Name:
          MAP-kinase activating death domain
          Gene Aliases:
          9630059K23Rik, IG20
          Chromosome Location:
          Chr.2: 91137360 - 91183991 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01271763_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Madd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177719.1 NP_001171190.1 22 - 23 3850 60
          NM_001177720.1 NP_001171191.1 20 - 21 3538 60
          NM_145527.4 NP_663502.3 21 - 22 3790 60
          GenBank mRNA AK122257.1 - 22 - 23 3692 60
          AK163518.1 - 4 - 5 484 60
          AY263980.1 - 22 - 23 3668 60
          AY263981.1 - 21 - 22 3614 60
          AY263982.1 - 20 - 21 3554 60
          AY263983.1 - 21 - 22 3557 60
          AY263984.1 - 20 - 21 3497 60
          AY263985.1 - 20 - 21 3479 60
          AY263986.1 - 20 - 21 3479 60
          AY263987.1 - 20 - 21 3479 60
          BC042212.1 - 11 - 12 1528 60
          BC063386.1 - 21 - 22 3591 5401819 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004108.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Madd

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          apoptotic process
          regulation of Rab protein signal transduction
          regulation of apoptotic process
          positive regulation of GTPase activity
          regulation of cell cycle
          negative regulation of growth hormone secretion
          execution phase of apoptosis
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of pancreatic amylase secretion
          negative regulation of apoptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway
          guanyl-nucleotide exchange factor activity
          Rab guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200034337.mm.1 NM_001177719.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034337.mm.1 NM_001177719.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034337.mm.1 NM_001177720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034337.mm.1 NM_001177720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034337.mm.1 NM_145527.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034337.mm.1 NM_145527.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034337.mm.1 TC0200004108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034341.mm.1 NM_001177719.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034341.mm.1 NM_001177719.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034341.mm.1 NM_001177720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034341.mm.1 NM_001177720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034341.mm.1 NM_145527.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034341.mm.1 NM_145527.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034341.mm.1 TC0200004108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034343.mm.1 NM_001177719.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034343.mm.1 NM_001177719.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034343.mm.1 NM_001177720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034343.mm.1 NM_001177720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034343.mm.1 NM_145527.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034343.mm.1 NM_145527.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034343.mm.1 TC0200004108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034342.mm.1 NM_001177719.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034342.mm.1 NM_001177719.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034342.mm.1 NM_001177720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034342.mm.1 NM_001177720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034342.mm.1 NM_145527.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034342.mm.1 NM_145527.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034342.mm.1 TC0200004108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034344.mm.1 NM_001177719.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034344.mm.1 NM_001177719.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034344.mm.1 NM_001177720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034344.mm.1 NM_001177720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034344.mm.1 NM_145527.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034344.mm.1 NM_145527.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034344.mm.1 TC0200004108.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline