Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01272189_g1
          See other GALNT5 GE Assays ›
          Gene Symbol:
          Galnt5
          Gene Name:
          UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 5
          Gene Aliases:
          4832424J23
          Chromosome Location:
          Chr.2: 57997264 - 58041027 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01272189_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Galnt5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172855.3 NP_766443.2 4 - 5 2084 91
          GenBank mRNA AK029323.1 - 2356 91
          BC116331.1 - 4 - 5 2083 91
          BC116332.1 - 4 - 5 2084 40096906 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000872.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Galnt5

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Process(es)

          alpha-1,6-mannosyltransferase activity
          alpha-1,2-mannosyltransferase activity
          mannosyltransferase activity
          alpha-1,3-mannosyltransferase activity
          alpha-1,3-galactosyltransferase activity
          UDP-glucose:glycoprotein glucosyltransferase activity
          glycolipid mannosyltransferase activity
          oligosaccharyl transferase activity
          dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
          polypeptide N-acetylgalactosaminyltransferase activity
          acetylglucosaminyltransferase activity
          acetylgalactosaminyltransferase activity
          galactosyltransferase activity
          fucosyltransferase activity
          O antigen polymerase activity
          lipopolysaccharide-1,6-galactosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          cellulose synthase activity
          9-phenanthrol UDP-glucuronosyltransferase activity
          1-phenanthrol glycosyltransferase activity
          9-phenanthrol glycosyltransferase activity
          1,2-dihydroxy-phenanthrene glycosyltransferase activity
          phenanthrol glycosyltransferase activity
          beta-1,4-mannosyltransferase activity
          carbohydrate binding
          alpha-1,2-galactosyltransferase activity
          dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
          lipopolysaccharide-1,5-galactosyltransferase activity
          dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          inositol phosphoceramide synthase activity
          glucosyltransferase activity
          metal ion binding
          alpha-(1->3)-fucosyltransferase activity
          alpha-(1->6)-fucosyltransferase activity
          indole-3-butyrate beta-glucosyltransferase activity
          salicylic acid glucosyltransferase (ester-forming) activity
          salicylic acid glucosyltransferase (glucoside-forming) activity
          benzoic acid glucosyltransferase activity
          chondroitin hydrolase activity
          dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
          cytokinin 9-beta-glucosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200007046.mm.1 NM_172855.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007046.mm.1 NM_172855.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007046.mm.1 TC0200000872.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007044.mm.1 NM_172855.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007044.mm.1 NM_172855.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007044.mm.1 TC0200000872.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007045.mm.1 NM_172855.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007045.mm.1 NM_172855.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007045.mm.1 TC0200000872.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0