Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01272320_m1
          See other 4930519A11RIK GE Assays ›
          Gene Symbol:
          4930519A11Rik
          Gene Name:
          RIKEN cDNA 4930519A11 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01272320_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930519A11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015836.1 - 1 - 2 206 64

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2214625 AK015836.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15280 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125813 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125813 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130545 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09871035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10415483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10361831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09913093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09945697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09930878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10183183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09945701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09841033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09913092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10426671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10216824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09931615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09779023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28246090 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432041_PM_at - -
          Mouse Genome 430 2.0 Array 1432041_at - -
          Mouse Genome 430 2.0 Array 1432041_PM_at - -
          Mouse Expression Set 430 1432041_at - -
          Mouse Expression Set 430 1432041_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432041_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432041_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 513934 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline