Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01273161_m1
          See other 9230104L09RIK GE Assays ›
          Gene Symbol:
          9230104L09Rik
          Gene Name:
          RIKEN cDNA 9230104L09 gene
          Gene Aliases:
          mCST E2
          Chromosome Location:
          Chr.2: 148844620 - 148850962 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01273161_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9230104L09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029960.3 NP_084236.1 2 - 3 373 57
          GenBank mRNA AK020314.1 - 2 - 3 374 57
          BC104302.1 - 2 - 3 345 40043696 57
          BC104303.1 - 2 - 3 345 40043697 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004815.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 9230104L09Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144894 NM_029960.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144894 AK020314.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144894 BC104303.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144894 NM_029960.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144894 AK020314.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144894 BC104303.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69919 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105171 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144894 NM_029960.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144894 AK020314.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144894 BC104303.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144894 NM_029960.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144894 AK020314.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144894 BC104303.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732527 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105171 - -
          Whole Mouse Genome G4122A A_51_P144894 NM_029960.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144894 AK020314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144894 BC104303.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144894 NM_029960.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144894 AK020314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144894 BC104303.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105171 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02245487 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02245494 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02245491 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02245499 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02245495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02178100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01926870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01926884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01926875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01926868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02136204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02245491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02178105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02216732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01926874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02178109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02178096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02216730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01926863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01926871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02136198 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10488412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488411 - -
          Affymetrix HT MG-430 PM Array Plate 1432531_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10488409 - -
          Mouse Genome 430 2.0 Array 1432531_PM_at - -
          Mouse Genome 430 2.0 Array 1432531_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77705_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77705_at - -
          Affymetrix Mouse Clariom S Array PSR0200041099.mm.1 NM_029960.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041099.mm.1 NM_029960.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041099.mm.1 TC0200004815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041104.mm.1 NM_029960.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041104.mm.1 NM_029960.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041104.mm.1 TC0200004815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041102.mm.1 NM_029960.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041102.mm.1 NM_029960.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041102.mm.1 TC0200004815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041100.mm.1 NM_029960.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041100.mm.1 NM_029960.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041100.mm.1 TC0200004815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041098.mm.1 NM_029960.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041098.mm.1 NM_029960.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041098.mm.1 TC0200004815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041103.mm.1 NM_029960.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041103.mm.1 NM_029960.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041103.mm.1 TC0200004815.mm.1 MTA TC
          Mouse Expression Set 430 1432531_PM_at - -
          Mouse Expression Set 430 1432531_at - -
          Affymetrix HT Mouse Genome 430B Array 1432531_at - -
          Affymetrix HT Mouse Genome 430B Array 1432531_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 580980 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 70324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2722414 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2722414 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2722414 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline