Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01273171_m1
          See other CST13 GE Assays ›
          Gene Symbol:
          Cst13
          Gene Name:
          cystatin 13
          Gene Aliases:
          1700006C19Rik, Cymg1, Mtcg1
          Chromosome Location:
          Chr.2: 148820099 - 148830410 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01273171_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cst13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027024.3 NP_081300.1 3 - 4 610 136
          GenBank mRNA AK005665.1 - 3 - 4 588 136
          AY600990.2 - 3 - 4 628 136
          BC048681.1 - 3 - 4 610 6774445 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002202.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cst13

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          negative regulation of peptidase activity
          molecular_function
          cysteine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491426 AK005665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491426 AY600990.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491426 BC048681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491426 NM_027024.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491426 AK005665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491426 AY600990.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491426 BC048681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491426 NM_027024.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18507 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102543 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161411 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162405 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173513 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491426 AK005665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491426 AY600990.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491426 BC048681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491426 NM_027024.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491426 AK005665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491426 AY600990.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491426 BC048681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491426 NM_027024.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732381 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173513 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162405 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161411 - -
          Whole Mouse Genome G4122A A_51_P491426 AK005665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491426 AY600990.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491426 BC048681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491426 NM_027024.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491426 AK005665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491426 AY600990.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491426 BC048681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491426 NM_027024.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136023 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162405 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173513 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161411 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949326 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01582672 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949325 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02187126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01949325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01899539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01651707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02197132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01840616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01255204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01450033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02050663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01840617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01705331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01899533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02006892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01849990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01606624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01949321 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10476923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476920 - -
          Affymetrix HT MG-430 PM Array Plate 1429424_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10476919 - -
          Mouse Genome 430 2.0 Array 1429424_at - -
          Mouse Genome 430 2.0 Array 1429424_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69294_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69294_at - -
          Affymetrix Mouse Clariom S Array PSR0200017427.mm.1 NM_027024.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017427.mm.1 NM_027024.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017427.mm.1 TC0200002202.mm.1 MTA TC
          Mouse Expression Set 430 1429424_PM_at - -
          Mouse Expression Set 430 1429424_at - -
          Murine Genome U74v2 Set 165487_f_at - -
          Murine Genome U74v2 Set 165487_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1429424_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429424_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 614659 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2595725 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2595725 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline