Search
Search

Zfp414| Gene Symbol: Zfp414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026712.3 | NP_080988.1 | 4 - 5 | 624 | 88 | |
| NR_034153.1 | 5 - 6 | 701 | 88 | |||
| GenBank mRNA | AK002706.1 | - | 4 - 5 | 614 | 88 | |
| AK169869.1 | - | 5 - 6 | 692 | 88 | ||
| BC038913.1 | - | 5 - 6 | 657 | 5099582 | 88 | |
| BC116933.1 | - | 4 - 5 | 597 | 40126252 | 88 | |
| BC119044.1 | - | 4 - 5 | 597 | 8733793 | 88 | |
| CX218166.1 | - | 5 - 6 | 689 | 88 | ||
Set Membership: |
|