Search
Search
| Gene Symbol: Nfya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110832.1 | NP_001104302.1 | 4 - 5 | 566 | 61 | |
| NM_010913.2 | NP_035043.1 | 3 - 4 | 479 | 61 | ||
| GenBank mRNA | AK004729.1 | - | 4 - 5 | 512 | 61 | |
| AK143723.1 | - | 3 - 4 | 441 | 61 | ||
| AK156487.1 | - | 4 - 5 | 396 | 61 | ||
| BC029695.1 | - | 3 - 4 | 367 | 5374597 | 61 | |
| BC057099.1 | - | 4 - 5 | 468 | 5716111 | 61 | |
| M86214.1 | - | 2 - 3 | 216 | 61 | ||
| M86215.1 | - | 3 - 4 | 300 | 61 | ||
| X55315.1 | - | 3 - 4 | 389 | 61 | ||
Set Membership: |
|