Search
Search
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010833.2 | NP_001010833.2 | 7 - 8 | 2290 | 60 | |
| GenBank mRNA | AK033009.1 | - | 7 - 8 | 2127 | 60 | |
| BC085140.1 | - | 2 - 3 | 159 | 60 | ||
| BC094363.1 | - | 3 - 4 | 993 | 60 | ||
Set Membership: |
|