Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01273967_m1
          See other THRSP GE Assays ›
          Gene Symbol:
          Thrsp
          Gene Name:
          thyroid hormone responsive
          Gene Aliases:
          S14
          Chromosome Location:
          Chr.7: 97412957 - 97417510 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01273967_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thrsp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009381.2 NP_033407.1 1 - 2 483 115
          GenBank mRNA AK077300.1 - 1 - 2 276 115
          BC009165.1 - 1 - 2 483 2811774 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003764.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thrsp

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          lipid metabolic process
          regulation of triglyceride biosynthetic process
          regulation of lipid biosynthetic process
          protein binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018666 AK077300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018666 BC009165.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018666 NM_009381.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424549 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174809 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018666 AK077300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018666 BC009165.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018666 NM_009381.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350430 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174809 - -
          Whole Mouse Genome G4122A A_51_P194099 BC009165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P194099 NM_009381.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103143 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111462 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174809 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975622 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975635 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975625 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06375697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06192677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06486748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06407368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05933802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05933804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06515217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06646681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06640734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05933803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06038926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06608166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05936058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350373 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10565610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565611 - -
          Affymetrix HT MG-430 PM Array Plate 1424737_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1444166_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422973_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10565609 - -
          Mouse Genome 430 2.0 Array 1444166_at - -
          Mouse Genome 430 2.0 Array 1422973_a_at - -
          Mouse Genome 430 2.0 Array 1444166_PM_at - -
          Mouse Genome 430 2.0 Array 1422973_PM_a_at - -
          Mouse Genome 430 2.0 Array 1424737_at - -
          Mouse Genome 430 2.0 Array 1424737_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21835_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21835_at - -
          Affymetrix Mouse Clariom S Array PSR0700031659.mm.1 NM_009381.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031659.mm.1 TC0700003764.mm.1 MTA TC
          Mouse Expression Set 430 1422973_a_at - -
          Mouse Expression Set 430 1424737_at - -
          Mouse Expression Set 430 1422973_PM_a_at - -
          Mouse Expression Set 430 1424737_PM_at - -
          Mouse Expression Set 430 1444166_PM_at - -
          Mouse Expression Set 430 1444166_at - -
          Murine Genome U74v2 Set 160306_PM_at - -
          Murine Genome U74v2 Set 160306_at - -
          Mouse Genome 430A 2.0 Array 1422973_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1424737_at - -
          Mouse Genome 430A 2.0 Array 1422973_a_at - -
          Mouse Genome 430A 2.0 Array 1424737_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424737_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422973_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422973_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424737_at - -
          Affymetrix HT Mouse Genome 430B Array 1444166_at - -
          Affymetrix HT Mouse Genome 430B Array 1444166_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 838610 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6980142 - -
          Illumina mouse-6 v1.0 expression beadchip 102690609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256775 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256775 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243094 - -
          mouse-6 v1.1 (Illumina) scl31033.2.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980142 - -
          Illumina Sentrix mouse-6.v1 bead array 102690609 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31033.2.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980142 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102690609 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256775 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256775 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243094 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31033.2.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline