Search
Search

Fam168a| Gene Symbol: Fam168a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178764.3 | NP_848879.1 | 3 - 4 | 318 | 65 | |
| GenBank mRNA | AK043083.1 | - | 2 - 3 | 297 | 65 | |
| AK043356.1 | - | 2 - 3 | 261 | 65 | ||
| AK046944.1 | - | 2 - 3 | 298 | 65 | ||
| AK047153.1 | - | 2 - 3 | 261 | 65 | ||
| AK048559.1 | - | 2 - 3 | 261 | 65 | ||
| AK051453.1 | - | 2 - 3 | 261 | 65 | ||
| AK084931.1 | - | 2 - 3 | 210 | 65 | ||
| AK122235.1 | - | 2 - 3 | 329 | 65 | ||
| AK137424.1 | - | 2 - 3 | 176 | 65 | ||
| AK162512.1 | - | 2 - 3 | 318 | 65 | ||
| BC076580.1 | - | 2 - 3 | 291 | 30542683 | 65 | |
| BC079886.1 | - | 3 - 4 | 318 | 6855858 | 65 | |
Set Membership: |
|