Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01274662_s1
          See other OLFR594 GE Assays ›
          Gene Symbol:
          Olfr594
          Gene Name:
          olfactory receptor 594
          Gene Aliases:
          MOR32-10
          Chromosome Location:
          Chr.7: 103219720 - 103220655 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01274662_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr594
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207143.1 NP_997026.1 1 672 58
          GenBank mRNA BC141016.1 - 1 749 9055628 58

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr594

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P216125 NM_207143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P216125 NM_207143.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654605 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142278 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P216125 NM_207143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P216125 NM_207143.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P216125 NM_207143.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P216125 NM_207143.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142278 - -
          Whole Mouse Genome G4122A A_51_P216125 NM_207143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216125 NM_207143.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138241 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05973985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06025755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05973612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05902556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05902559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06141504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06141505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382528 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555776 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555775 - -
          Affymetrix Mouse Clariom S Array PSR0700012737.mm.1 TC0700001485.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012735.mm.1 TC0700001485.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012736.mm.1 TC0700001485.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012736.mm.1 NM_207143.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012736.mm.1 NM_207143.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 616270 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3450088 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450088 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226190 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226190 - -
          mouse-6 v1.1 (Illumina) scl0258246.1_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 3450088 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258246.1_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450088 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226190 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226190 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline