Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01274942_s1
          See other OLFR517 GE Assays ›
          Gene Symbol:
          Olfr517
          Gene Name:
          olfactory receptor 517
          Gene Aliases:
          MOR268-4
          Chromosome Location:
          Chr.7: 108868208 - 108869152 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01274942_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr517
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011846.1 NP_001011846.1 1 639 70
          GenBank mRNA BC150830.1 - 1 662 9087741 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr517

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125458 NM_001011846.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125458 BC150830.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P125458 NM_001011846.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P125458 BC150830.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404799 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06187495 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06187486 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06187496 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06187488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06187497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05987379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06413226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06187494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06187496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06108101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06187502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06500565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06413239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06579686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06289978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05987373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404842 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566703 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566702 - -
          Affymetrix Mouse Clariom S Array PSR0700033561.mm.1 TC0700004062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033562.mm.1 NM_001011846.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033562.mm.1 NM_001011846.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033562.mm.1 TC0700004062.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 865145 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2913656 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2913656 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline