Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01275230_m1
          See other CDSN GE Assays ›
          Gene Symbol:
          Cdsn
          Gene Name:
          corneodesmosin
          Gene Aliases:
          AI747712
          Chromosome Location:
          Chr.17: 35543082 - 35557180 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01275230_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdsn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008424.2 NP_001008424.2 1 - 2 159 58
          GenBank mRNA BC055373.1 - 1 - 2 156 30058959 58
          BY771914.1 - 1 - 2 115 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000694.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdsn

          Gene Ontology Categories:

          Function(s) Process(es)

          single organismal cell-cell adhesion
          skin morphogenesis
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2190994 AK133029.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2190994 BC055373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2190994 NM_001008424.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81704 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142266 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2190994 AK133029.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2190994 BC055373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2190994 NM_001008424.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159700 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142266 - -
          Whole Mouse Genome G4122A A_52_P569998 BC055373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P569998 NM_001008424.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142266 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13455679 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13072525 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13072523 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13072524 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13455678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13314936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13455675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13314943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13555421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13429895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13429909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13189529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13555422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13090447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13380806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13479217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13429889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13376552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13455678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13193525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159648 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444885 - -
          Affymetrix HT MG-430 PM Array Plate 1435191_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444883 - -
          Mouse Genome 430 2.0 Array 1435191_at - -
          Mouse Genome 430 2.0 Array 1444607_at - -
          Mouse Genome 430 2.0 Array 1435191_PM_at - -
          Mouse Genome 430 2.0 Array 1444607_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 386463_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 386463_at - -
          Affymetrix Mouse Clariom S Array PSR1700007134.mm.1 NM_001008424.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007134.mm.1 NM_001008424.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007134.mm.1 TC1700000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007135.mm.1 NM_001008424.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007135.mm.1 NM_001008424.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007135.mm.1 TC1700000694.mm.1 MTA TC
          Mouse Expression Set 430 1435191_PM_at - -
          Mouse Expression Set 430 1435191_at - -
          Murine Genome U74v2 Set 104363_at - -
          Murine Genome U74v2 Set 104363_PM_at - -
          Mouse Genome 430A 2.0 Array 1435191_at - -
          Mouse Genome 430A 2.0 Array 1435191_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435191_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435191_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253772 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235732 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2796382 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2796382 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline