Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01275749_g1
          See other IMMP1L GE Assays ›
          Gene Symbol:
          Immp1l
          Gene Name:
          IMP1 inner mitochondrial membrane peptidase-like (S. cerevisiae)
          Gene Aliases:
          1500034J20Rik, 2610528O17Rik, AU042386
          Chromosome Location:
          Chr.2: 105904638 - 105965558 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01275749_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Immp1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028260.2 NP_082536.1 3 - 4 337 98
          GenBank mRNA AK005356.1 - 3 - 4 338 98
          AK009570.1 - 3 - 4 295 98
          AK010075.1 - 3 - 4 295 98
          AK011278.1 - 3 - 4 338 98
          AK011382.1 - 3 - 4 338 98
          AK011420.1 - 3 - 4 299 98
          AK011466.1 - 3 - 4 310 98
          AK011618.1 - 3 - 4 299 98
          AK011872.1 - 3 - 4 301 98
          AK012181.1 - 290 98
          AK015978.1 - 3 - 4 308 98
          AK078300.1 - 3 - 4 324 98
          AK135670.1 - 3 - 4 297 98
          AK168202.1 - 3 - 4 286 98
          BC008259.1 - 3 - 4 296 2649406 98
          BC081433.1 - 2 - 3 180 6814816 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Immp1l

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein processing involved in protein targeting to mitochondrion
          biological_process
          molecular_function
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200011923.mm.1 NM_028260.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011923.mm.1 NM_028260.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011923.mm.1 TC0200001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011926.mm.1 NM_028260.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011926.mm.1 NM_028260.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011926.mm.1 TC0200001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011928.mm.1 NM_028260.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011928.mm.1 NM_028260.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011928.mm.1 TC0200001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011927.mm.1 NM_028260.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011927.mm.1 NM_028260.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011927.mm.1 TC0200001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011924.mm.1 NM_028260.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011924.mm.1 NM_028260.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011924.mm.1 TC0200001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011925.mm.1 NM_028260.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011925.mm.1 NM_028260.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011925.mm.1 TC0200001586.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 900524 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline