Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01275871_m1
          See other 1700026D11RIK GE Assays ›
          Gene Symbol:
          1700026D11Rik
          Gene Name:
          RIKEN cDNA 1700026D11 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 132490728 - 132524057 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01275871_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700026D11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028286.1 2 - 3 380 68
          GenBank mRNA AK006376.1 - 2 - 3 381 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002034.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214646 AK006376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214646 NR_028286.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214646 AK006376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214646 NR_028286.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295186 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906717 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16945 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141441 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214646 AK006376.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214646 NR_028286.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214646 AK006376.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214646 NR_028286.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04697206 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04697189 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141441 - -
          Whole Mouse Genome G4122A A_52_P214646 AK006376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214646 NR_028286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214646 AK006376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214646 NR_028286.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124170 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01969744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640627 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200016199.mm.1 NR_028286.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016199.mm.1 NR_028286.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016199.mm.1 TC0200002034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016211.mm.1 NR_028286.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016211.mm.1 NR_028286.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016211.mm.1 TC0200002034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016212.mm.1 NR_028286.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016212.mm.1 NR_028286.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016212.mm.1 TC0200002034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016194.mm.1 NR_028286.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016194.mm.1 NR_028286.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016194.mm.1 TC0200002034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016193.mm.1 NR_028286.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016193.mm.1 NR_028286.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016193.mm.1 TC0200002034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016213.mm.1 NR_028286.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016213.mm.1 NR_028286.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016213.mm.1 TC0200002034.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline