Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01276061_s1
          See other OLFR1222 GE Assays ›
          Gene Symbol:
          Olfr1222
          Gene Name:
          olfactory receptor 1222
          Gene Aliases:
          MOR233-14
          Chromosome Location:
          Chr.2: 89124794 - 89125729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01276061_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1222
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011860.1 NP_001011860.1 1 889 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1222

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312024 NM_001011860.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312024 NM_001011860.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715740 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312024 NM_001011860.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312024 NM_001011860.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312024 NM_001011860.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312024 NM_001011860.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00427298 - -
          Whole Mouse Genome G4122A A_51_P312024 NM_001011860.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312024 NM_001011860.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02064559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02202153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412699 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484792 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484791 - -
          Affymetrix Mouse Clariom S Array PSR0200034014.mm.1 NM_001011860.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034014.mm.1 NM_001011860.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034014.mm.1 TC0200004043.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034013.mm.1 NM_001011860.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034013.mm.1 NM_001011860.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034013.mm.1 TC0200004043.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 876057 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161191 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161191 - -
          mouse-6 v1.1 (Illumina) GI_58801477-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161191 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161191 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0