Search
Search

Prss53| Gene Symbol: Prss53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081268.1 | NP_001074737.1 | 5 - 6 | 717 | 91 | |
| GenBank mRNA | AK220244.1 | - | 5 - 6 | 674 | 91 | |
| BC039632.1 | - | 2 - 3 | 287 | 91 | ||
| BC147509.1 | - | 5 - 6 | 714 | 9056457 | 91 | |
| BC151010.1 | - | 5 - 6 | 714 | 9087923 | 91 | |
| BC151017.1 | - | 5 - 6 | 714 | 9087930 | 91 | |
| BC172059.1 | - | 5 - 6 | 714 | 9054345 | 91 | |
Set Membership: |
|