Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01277542_m1
          See other 1700007K09RIK GE Assays ›
          Gene Symbol:
          1700007K09Rik
          Gene Name:
          RIKEN cDNA 1700007K09 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 131325930 - 131331434 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01277542_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700007K09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027037.2 NP_081313.1 3 - 4 282 128
          GenBank mRNA AK005730.1 - 3 - 4 277 128
          BC048596.1 - 3 - 4 282 6740370 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004395.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700007K09Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980677 NM_027037.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980677 AK005730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980677 BC048596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300395 NM_027037.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300395 AK005730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300395 BC048596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300395 NM_027037.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300395 AK005730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300395 BC048596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117966 NM_027037.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117966 AK005730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117966 BC048596.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532836 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120062 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980677 NM_027037.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980677 AK005730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980677 BC048596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117966 NM_027037.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117966 AK005730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117966 BC048596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300395 NM_027037.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300395 AK005730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300395 BC048596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300395 NM_027037.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300395 AK005730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300395 BC048596.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25527844 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120062 - -
          Whole Mouse Genome G4122A A_51_P300395 NM_027037.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300395 AK005730.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300395 BC048596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300395 NM_027037.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300395 AK005730.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300395 BC048596.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06501195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06534919 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06638384 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06535483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06638379 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06131867 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06131870 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06131865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06288996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06137980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25527836 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568516 - -
          Affymetrix HT MG-430 PM Array Plate 1430169_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568512 - -
          Mouse Genome 430 2.0 Array 1430169_at - -
          Mouse Genome 430 2.0 Array 1430169_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69318_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69318_at - -
          Affymetrix Mouse Clariom S Array PSR0700036993.mm.1 NM_027037.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036993.mm.1 NM_027037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036993.mm.1 TC0700004395.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036994.mm.1 NM_027037.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036994.mm.1 NM_027037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036994.mm.1 TC0700004395.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036992.mm.1 NM_027037.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036992.mm.1 NM_027037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036992.mm.1 TC0700004395.mm.1 MTA TC
          Mouse Expression Set 430 1430169_PM_at - -
          Mouse Expression Set 430 1430169_at - -
          Murine Genome U74v2 Set 168047_PM_at - -
          Murine Genome U74v2 Set 168047_at - -
          Affymetrix HT Mouse Genome 430B Array 1430169_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430169_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 856192 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6450402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2834950 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2834950 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline