Search
Search

Gm13629| Gene Symbol: Gm13629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033495.1 | 1 - 2 | 358 | 70 | ||
| GenBank mRNA | AK020721.1 | - | 1 - 2 | 360 | 70 | |
| AK039017.1 | - | 1 - 2 | 343 | 70 | ||
| AK045660.1 | - | 1 - 2 | 361 | 70 | ||
| AK138511.1 | - | 1 - 2 | 359 | 70 | ||
| AK138532.1 | - | 1 - 2 | 359 | 70 | ||
Set Membership: |
|