| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400001302.mm.1 |
NR_033571.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001302.mm.1 |
NR_033571.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001302.mm.1 |
TC1400000167.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400001307.mm.1 |
NR_033571.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001307.mm.1 |
NR_033571.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001307.mm.1 |
TC1400000167.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400001305.mm.1 |
NR_033571.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001305.mm.1 |
NR_033571.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001305.mm.1 |
TC1400000167.mm.1 |
MTA TC |