Search
Search

Trmt6| Gene Symbol: Trmt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175113.3 | NP_780322.2 | 6 - 7 | 790 | 73 | |
| GenBank mRNA | AK014359.1 | - | 6 - 7 | 906 | 73 | |
| AK017672.1 | - | 6 - 7 | 732 | 73 | ||
| AK028755.1 | - | 6 - 7 | 790 | 73 | ||
| AK129300.1 | - | 6 - 7 | 757 | 73 | ||
| AK164396.1 | - | 6 - 7 | 723 | 73 | ||
| AK165976.1 | - | 6 - 7 | 742 | 73 | ||
| AK167222.1 | - | 6 - 7 | 756 | 73 | ||
| BC049083.1 | - | 3 - 4 | 547 | 73 | ||
| BC052648.1 | - | 6 - 7 | 708 | 30030801 | 73 | |
Set Membership: |
|