Search
Search

Gsg1l| Gene Symbol: Gsg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101488.1 | NP_001094958.1 | 3 - 4 | 751 | 59 | |
| GenBank mRNA | AK082731.1 | - | 3 - 4 | 753 | 59 | |
| BM950164.1 | - | 1 - 2 | 141 | 59 | ||
Set Membership: |
|