Search
Search

Mki67| Gene Symbol: Mki67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081117.2 | NP_001074586.2 | 7 - 8 | 1776 | 73 | |
| GenBank mRNA | AK052772.1 | - | 7 - 8 | 1696 | 73 | |
| AK143677.1 | - | 7 - 8 | 1815 | 73 | ||
| X82786.1 | - | 7 - 8 | 1884 | 73 | ||
Set Membership: |
|