Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01278646_g1
          See other CNGA4 GE Assays ›
          Gene Symbol:
          Cnga4
          Gene Name:
          cyclic nucleotide gated channel alpha 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 105400377 - 105409313 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01278646_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnga4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033317.3 NP_001028489.1 5 - 6 1390 77
          GenBank mRNA AK136723.1 - 5 - 6 1392 77
          BC107347.1 - 5 - 6 1336 40055406 77
          BC107348.2 - 4 - 5 1515 40055413 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001551.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Ion Channel 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          sensory perception of smell
          ion transmembrane transport
          regulation of ion transmembrane transport
          regulation of membrane potential
          response to stimulus
          protein heterotetramerization
          transmembrane transport
          potassium ion transmembrane transport
          cation transmembrane transport
          nucleotide binding
          ion channel activity
          intracellular cyclic nucleotide activated cation channel activity
          intracellular cAMP activated cation channel activity
          intracellular cGMP activated cation channel activity
          voltage-gated potassium channel activity
          cAMP binding
          cGMP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019542 NM_001033317.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019542 AK136723.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73413 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019542 NM_001033317.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019542 AK136723.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25392157 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06310798 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06310798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25392118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06104423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06427392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06315073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06104413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06427388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05969765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25392177 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555929 - -
          Affymetrix HT MG-430 PM Array Plate 1445339_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10555925 - -
          Mouse Genome 430 2.0 Array 1445339_at - -
          Mouse Genome 430 2.0 Array 1445339_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233649_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233649_at - -
          Affymetrix Mouse Clariom S Array PSR0700012958.mm.1 TC0700001551.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012958.mm.1 NM_001033317.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012958.mm.1 NM_001033317.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012959.mm.1 TC0700001551.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012959.mm.1 NM_001033317.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012959.mm.1 NM_001033317.3 Refseq
          Mouse Expression Set 430 1445339_PM_at - -
          Mouse Expression Set 430 1445339_at - -
          Affymetrix HT Mouse Genome 430B Array 1445339_at - -
          Affymetrix HT Mouse Genome 430B Array 1445339_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 340472 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106450044 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222098 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2875896 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2875896 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline