Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01279003_g1
          See other LMNTD2 GE Assays ›
          Gene Symbol:
          Lmntd2
          Gene Name:
          lamin tail domain containing 2
          Gene Aliases:
          1600016N20Rik
          Chromosome Location:
          Chr.7: 141209990 - 141215195 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01279003_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lmntd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028050.2 NP_082326.1 11 - 12 1632 65
          GenBank mRNA AK005476.1 - 2 - 3 219 65
          BC119093.1 - 11 - 12 1636 8733842 65
          BC119119.1 - 11 - 12 1636 8733868 65
          BC144834.1 - 11 - 12 1666 9053382 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004526.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of mRNA splicing, via spliceosome
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25587465 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25587467 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 BC119093.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 BC119119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 BC144834.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 NM_028050.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 AK005476.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 BC119093.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 BC119119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 BC144834.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 NM_028050.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401069 AK005476.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773998 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 BC119093.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 BC119119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 BC144834.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 NM_028050.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 AK005476.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 BC119093.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 BC119119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 BC144834.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 NM_028050.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401069 AK005476.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06123615 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06123612 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06123577 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06123595 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25587465 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25587467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25587494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06522002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06123621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06522010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25587449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569093 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569086 - -
          Affymetrix HT MG-430 PM Array Plate 1430017_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569085 - -
          Mouse Genome 430 2.0 Array 1430017_PM_at - -
          Mouse Genome 430 2.0 Array 1430017_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72000_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72000_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700037995.mm.1 NM_028050.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037995.mm.1 NM_028050.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037995.mm.1 TC0700004526.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037994.mm.1 NM_028050.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037994.mm.1 NM_028050.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037994.mm.1 TC0700004526.mm.1 MTA TC
          Mouse Expression Set 430 1430017_PM_at - -
          Mouse Expression Set 430 1430017_at - -
          Affymetrix HT Mouse Genome 430B Array 1430017_at - -
          Affymetrix HT Mouse Genome 430B Array 1430017_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 828982 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5290519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242629 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242629 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242629 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline